Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
csnk-1
Entrez Gene ID
172891
Gene Name
Casein kinase I gamma
Gene Aliases
CELE_Y106G6E.6
Chromosome Location
Chr.I: 10226986 - 10233386 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060293.6 NP_492694.1 6 - 7 1150 59

View Full Product Details