Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dlc-6
Entrez Gene ID
190923
Gene Name
Dynein Light Chain
Gene Aliases
CELE_Y106G6G.3
Chromosome Location
Chr.I: 10323511 - 10324145 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060305.2 NP_492706.1 1 - 2 317 105

View Full Product Details