Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aph-2
Entrez Gene ID
172905
Gene Name
protein-coding
Gene Aliases
CELE_ZC434.6
Chromosome Location
Chr.I: 10340997 - 10345654 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129091.3 NP_001122563.1 4 - 5 1355 97
NM_060311.4 NP_492712.2 4 - 5 1355 97

View Full Product Details