Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tag-68
Entrez Gene ID
172930
Gene Name
Dwarfin sma
Gene Aliases
CELE_F37D6.6
Chromosome Location
Chr.I: 10501585 - 10505949 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060345.4 NP_492746.2 1 - 2 260 73

View Full Product Details