Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
che-13
Entrez Gene ID
186607
Gene Name
protein-coding
Gene Aliases
CELE_F59C6.7
Chromosome Location
Chr.I: 10506998 - 10515586 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001264468.3 NP_001251397.1 4 - 5 434 87
GenBank mRNA AF520567.1 - 4 - 5 354 87

View Full Product Details