Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bli-3
Entrez Gene ID
171608
Gene Name
Dual oxidase 1
Gene Aliases
CELE_F56C11.1
Chromosome Location
Chr.I: 146486 - 154855 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058285.5 NP_490686.3 13 - 14 3053 72

View Full Product Details