Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-11
Entrez Gene ID
171609
Gene Name
protein-coding
Gene Aliases
CELE_F56C11.2
Chromosome Location
Chr.I: 160172 - 169763 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306284.3 NP_001293213.1 7 - 8 1293 91
NM_058286.7 NP_490687.1 12 - 13 1864 91

View Full Product Details