Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atm-1
Entrez Gene ID
3565793
Gene Name
protein-coding
Gene Aliases
CELE_Y48G1BL.2
Chromosome Location
Chr.I: 183364 - 208468 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306285.2 NP_001293214.1 19 - 20 4909 93
NM_001306286.3 NP_001293215.1 10 - 11 2866 93

View Full Product Details