Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnp-8
Entrez Gene ID
171627
Gene Name
protein-coding
Gene Aliases
CELE_R119.7
Chromosome Location
Chr.I: 364510 - 368579 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383945.1 3 - 4 521 90
NM_058323.5 NP_490724.1 2 - 3 621 90

View Full Product Details