Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
deps-1
Entrez Gene ID
171642
Gene Name
protein-coding
Gene Aliases
CELE_Y65B4BL.2
Chromosome Location
Chr.I: 506473 - 509056 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306264.3 NP_001293193.1 6 - 7 983 91
NM_058341.6 NP_490742.1 7 - 8 1122 91

View Full Product Details