Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gpa-16
Entrez Gene ID
171675
Gene Name
protein-coding
Gene Aliases
CELE_Y95B8A.5
Chromosome Location
Chr.I: 897286 - 907370 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306257.3 NP_001293186.1 2 - 3 258 91
NM_001306258.3 NP_001293187.1 1 - 2 225 91
NM_058389.4 NP_490790.1 7 - 8 955 91
GenBank mRNA AY008138.1 - 7 - 8 951 91

View Full Product Details