Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
haf-6
Entrez Gene ID
171694
Gene Name
protein-coding
Gene Aliases
CELE_Y48G8AL.11
Chromosome Location
Chr.I: 1162880 - 1175868 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058427.6 NP_490828.3 1 - 2 179 57

View Full Product Details