Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
afd-1
Entrez Gene ID
171776
Gene Name
protein-coding
Gene Aliases
CELE_W03F11.6
Chromosome Location
Chr.I: 2160038 - 2209644 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026489.7 NP_001021660.2 4 - 5 685 82
NM_001026490.6 NP_001021661.2 4 - 5 683 82
NM_001306400.4 NP_001293329.1 4 - 5 687 82
NM_058537.5 NP_490938.3 4 - 5 679 82

View Full Product Details