Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
abt-2
Entrez Gene ID
171782
Gene Name
protein-coding
Gene Aliases
CELE_F12B6.1
Chromosome Location
Chr.I: 2255921 - 2272866 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381367.1 NP_001368466.1 30 - 31 5477 86
NM_058548.7 NP_490949.3 30 - 31 5588 86

View Full Product Details