Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdk-7
Entrez Gene ID
171784
Gene Name
Cyclin-dependent kinase 7
Gene Aliases
CELE_Y39G10AL.3
Chromosome Location
Chr.I: 2281501 - 2291163 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383811.1 6 - 7 825 72
GenBank mRNA AF154004.1 - 6 - 7 756 72

View Full Product Details