Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zeel-1
Entrez Gene ID
189770
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y39G10AR.5
Chromosome Location
Chr.I: 2342216 - 2350536 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058565.4 NP_490966.2 12 - 13 2524 141

View Full Product Details