Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zeel-1
Entrez Gene ID
189770
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y39G10AR.5
Chromosome Location
Chr.I: 2342216 - 2350536 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058565.4 NP_490966.2 2 - 3 375 130

View Full Product Details