Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nekl-1
Entrez Gene ID
189769
Gene Name
protein-coding
Gene Aliases
CELE_Y39G10AR.3
Chromosome Location
Chr.I: 2357883 - 2372985 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306391.3 NP_001293320.1 16 - 17 2226 74
NM_001306392.3 NP_001293321.1 15 - 16 2112 74
NR_131355.1 16 - 17 2226 74
NR_131356.1 15 - 16 2115 74

View Full Product Details