Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zwl-1
Entrez Gene ID
171798
Gene Name
protein-coding
Gene Aliases
CELE_Y39G10AR.2
Chromosome Location
Chr.I: 2402230 - 2409999 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001262221.3 NP_001249150.1 3 - 4 470 110
NM_001262222.4 NP_001249151.1 3 - 4 472 110

View Full Product Details