Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dmsr-2
Entrez Gene ID
189527
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y23H5B.4
Chromosome Location
Chr.I: 2533268 - 2542114 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058591.6 NP_490992.2 3 - 4 704 72

View Full Product Details