Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbp-1
Entrez Gene ID
266822
Gene Name
FBPase domain-containing protein
Gene Aliases
CELE_K07A3.1
Chromosome Location
Chr.I: 2580935 - 2587110 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058603.7 NP_491004.1 3 - 4 707 69
NR_131360.1 2 - 3 577 69
GenBank mRNA AJ271466.1 - 2 - 3 581 69

View Full Product Details