Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccb-2
Entrez Gene ID
189330
Gene Name
protein-coding
Gene Aliases
CELE_W10C8.1
Chromosome Location
Chr.I: 2845859 - 2849903 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306451.2 NP_001293380.1 5 - 6 949 96
NM_001306452.1 NP_001293381.1 5 - 6 949 96
NR_131378.1 5 - 6 1132 96
NR_131379.1 5 - 6 992 96

View Full Product Details