Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fog-1
Entrez Gene ID
171895
Gene Name
protein-coding
Gene Aliases
CELE_Y54E10A.4
Chromosome Location
Chr.I: 3211040 - 3217366 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381585.1 NP_001367957.1 4 - 5 529 69
NM_001381586.2 NP_001367956.1 4 - 5 535 69
GenBank mRNA AJ297846.1 - 4 - 5 551 69
AJ297847.1 - 4 - 5 551 69

View Full Product Details