Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smp-2
Entrez Gene ID
183905
Gene Name
Sema domain-containing protein
Gene Aliases
CELE_D1037.2
Chromosome Location
Chr.I: 3666487 - 3671700 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058796.5 NP_491197.3 9 - 10 1703 71

View Full Product Details