Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- asic-1
- Entrez Gene ID
-
191422
- Gene Name
- Degenerin-like protein asic-1
- Gene Aliases
-
CELE_ZK770.1
- Chromosome Location
-
Chr.I: 3769672 - 3774005 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058813.5 |
NP_491214.3 |
10 - 11 |
1521 |
68 |
- Gene Symbol
- C32E8.6
- Entrez Gene ID
-
171945
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C32E8.6
- Chromosome Location
-
Chr.I: 3769662 - 3777468 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001384008.1 |
|
10 - 11 |
1529 |
68 |
View Full Product Details