Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pbs-4
Entrez Gene ID
171975
Gene Name
Proteasome subunit beta type-2
Gene Aliases
CELE_T20F5.2
Chromosome Location
Chr.I: 3908747 - 3909701 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058860.7 NP_491261.1 3 - 4 475 73

View Full Product Details