Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lrp-2
Entrez Gene ID
171981
Gene Name
Low-density lipoprotein RecePtor related
Gene Aliases
CELE_T21E3.3
Chromosome Location
Chr.I: 3935272 - 3953991 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058869.5 NP_491270.2 11 - 12 7819 66

View Full Product Details