Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpn-3
Entrez Gene ID
171986
Gene Name
Calponin-homology (CH) domain-containing protein
Gene Aliases
CELE_F28H1.2
Chromosome Location
Chr.I: 3987842 - 3988699 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058881.6 NP_491282.1 2 - 3 327 76

View Full Product Details