Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhc-1
Entrez Gene ID
172041
Gene Name
protein-coding
Gene Aliases
CELE_T21E12.4
Chromosome Location
Chr.I: 4386777 - 4401907 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058962.6 NP_491363.1 1 - 2 679 83

View Full Product Details