Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-17
Entrez Gene ID
172059
Gene Name
Homeobox domain-containing protein
Gene Aliases
CELE_D1007.1
Chromosome Location
Chr.I: 4568760 - 4570027 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058992.5 NP_491393.1 1 - 2 166 124
GenBank mRNA AF272397.1 - 1 - 2 186 124

View Full Product Details