Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fncm-1
Entrez Gene ID
172159
Gene Name
FANCM (Fanconi anemia complex component M) homolog
Gene Aliases
CELE_F56A3.1
Chromosome Location
Chr.I: 5190138 - 5195243 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059142.4 NP_491543.1 3 - 4 523 79

View Full Product Details