Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- spe-11
- Entrez Gene ID
-
172179
- Gene Name
- Spermatocyte protein spe-11
- Gene Aliases
-
CELE_F48C1.7
- Chromosome Location
-
Chr.I: 5330578 - 5332194 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059168.4 |
NP_491569.1 |
3 - 4 |
513 |
109 |
- Gene Symbol
- F48C1.9
- Entrez Gene ID
-
266832
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F48C1.9
- Chromosome Location
-
Chr.I: 5330525 - 5332780 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383075.1 |
|
4 - 5 |
891 |
109 |
View Full Product Details