Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk-1
Entrez Gene ID
172184
Gene Name
PIPK domain-containing protein
Gene Aliases
CELE_F55A12.3
Chromosome Location
Chr.I: 5358522 - 5392671 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059175.6 NP_491576.2 5 - 6 489 132

View Full Product Details