Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frm-4
Entrez Gene ID
172189
Gene Name
protein-coding
Gene Aliases
CELE_C24A11.8
Chromosome Location
Chr.I: 5402882 - 5408344 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001262958.3 NP_001249887.1 3 - 4 245 140
NM_001383090.1 3 - 4 247 140
NM_059185.7 NP_491586.1 3 - 4 247 116

View Full Product Details