Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-10
Entrez Gene ID
172229
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_F55F8.1
Chromosome Location
Chr.I: 5662284 - 5667160 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383129.1 5 - 6 694 119

View Full Product Details