Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtk-1
Entrez Gene ID
172246
Gene Name
protein-coding
Gene Aliases
CELE_B0414.7
Chromosome Location
Chr.I: 5795400 - 5803734 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380377.2 NP_001367356.1 9 - 10 2662 80
NM_059282.5 NP_491683.1 10 - 11 2714 80

View Full Product Details