Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-4
Entrez Gene ID
181982
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_B0414.1
Chromosome Location
Chr.I: 5801872 - 5803294 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374897.2 NP_001361914.1 2 - 3 264 153

View Full Product Details