The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_059309.8 | NP_491710.2 | 3 - 4 | 358 | 125 |
NM_182013.8 | NP_871813.1 | 2 - 3 | 302 | 125 |