Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rtel-1
- Entrez Gene ID
-
172946
- Gene Name
- Regulator of telomere elongation helicase 1 homolog
- Gene Aliases
-
CELE_F25H2.13
- Chromosome Location
-
Chr.I: 10571954 - 10577135 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_060368.6 |
NP_492769.1 |
5 - 6 |
1325 |
143 |
- Gene Symbol
- F29C6.1
- Entrez Gene ID
-
172947
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F29C6.1
- Chromosome Location
-
Chr.I: 10572023 - 10580960 on Build WBcel235
View Full Product Details