Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pxf-1
Entrez Gene ID
177894
Gene Name
protein-coding
Gene Aliases
CELE_T14G10.2
Chromosome Location
Chr.IV: 10164288 - 10174341 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028218.6 NP_001023389.1 6 - 7 1855 108
NM_001028219.3 NP_001023390.1 6 - 7 1855 108
NM_001268536.3 NP_001255465.1 2 - 3 1475 108
NM_001268537.3 NP_001255466.1 1 - 2 1349 108
NM_001380435.1 NP_001367298.1 2 - 3 1475 108
NM_001380436.1 NP_001367299.1 1 - 2 1349 108
GenBank mRNA AF170796.1 - 6 - 7 1844 108
AF308447.1 - 6 - 7 1844 108
AF308448.1 - 6 - 7 1844 108
AF308449.1 - 3 - 4 1542 108

View Full Product Details