Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gon-14
Entrez Gene ID
179052
Gene Name
protein-coding
Gene Aliases
CELE_F44C4.4
Chromosome Location
Chr.V: 6606406 - 6611158 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171475.5 NP_741557.2 12 - 13 2160 77
NM_171476.5 NP_741558.2 12 - 13 2160 77

View Full Product Details