Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mod-1
Entrez Gene ID
179269
Gene Name
protein-coding
Gene Aliases
CELE_K06C4.6
Chromosome Location
Chr.V: 8909704 - 8914040 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028863.5 NP_001024034.1 4 - 5 483 93
NM_001028864.4 NP_001024035.1 4 - 5 435 93
NM_171495.6 NP_741580.1 4 - 5 476 93
GenBank mRNA AF303088.1 - 4 - 5 435 93

View Full Product Details