Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spas-1
Entrez Gene ID
179300
Gene Name
protein-coding
Gene Aliases
CELE_C24B5.2
Chromosome Location
Chr.V: 9175789 - 9177997 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269186.3 NP_001256115.1 8 - 9 1451 67
NM_171501.6 NP_741586.1 7 - 8 1265 67
NR_136364.1 6 - 7 1630 67
GenBank mRNA AB287436.1 - 8 - 9 1437 67

View Full Product Details