Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
xbx-6
Entrez Gene ID
179361
Gene Name
protein-coding
Gene Aliases
CELE_F40F9.1
Chromosome Location
Chr.V: 9703055 - 9707835 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373079.3 NP_001359646.1 5 - 5 497 70
NM_171511.7 NP_741597.1 5 - 6 499 70
NR_069217.1 5 - 6 502 70

View Full Product Details