Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-2
Entrez Gene ID
179377
Gene Name
Seven TM Receptor
Gene Aliases
CELE_C50C10.7
Chromosome Location
Chr.V: 9822619 - 9824249 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171516.3 NP_741602.1 2 - 3 292 154
GenBank mRNA AF216966.1 - 2 - 3 292 154

View Full Product Details