Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lsy-12
Entrez Gene ID
3565873
Gene Name
protein-coding
Gene Aliases
CELE_R07B5.9
Chromosome Location
Chr.V: 9846157 - 9859963 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276918.3 NP_001263847.1 14 - 15 4080 84
NM_001276919.3 NP_001263848.1 12 - 13 3228 84
NM_001276920.4 NP_001263849.1 15 - 16 4215 84
NM_001373081.2 NP_001360180.1 13 - 13 3430 84

View Full Product Details