Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dmsr-1
Entrez Gene ID
186437
Gene Name
protein-coding
Gene Aliases
CELE_F57B7.1
Chromosome Location
Chr.V: 11438764 - 11443450 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171541.6 NP_741629.1 5 - 6 1355 115

View Full Product Details