Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
max-1
Entrez Gene ID
179794
Gene Name
protein-coding
Gene Aliases
CELE_C34B4.1
Chromosome Location
Chr.V: 13098903 - 13120220 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380794.1 NP_001367137.1 6 - 7 1355 65
NM_001383435.1 7 - 8 1497 65
GenBank mRNA AY084080.1 - 6 - 7 1355 65

View Full Product Details