Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpt-5
Entrez Gene ID
179881
Gene Name
Carn_acyltransf domain-containing protein
Gene Aliases
CELE_F09F3.9
Chromosome Location
Chr.V: 13873195 - 13875891 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171549.5 NP_741637.1 1 - 2 90 87

View Full Product Details