Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-53
Entrez Gene ID
180053
Gene Name
Nuclear hormone receptor family member nhr-53
Gene Aliases
CELE_K06B4.11
Chromosome Location
Chr.V: 15700606 - 15702363 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171556.5 NP_741644.1 2 - 3 217 85

View Full Product Details